Få mere at vide
[((R,R)-2-Amino-1,2-diphenylethyl)[(4-tolyl)sulfonyl]amido](chloro)(η6-p-cymene)ruthenium, TRC
Artikelnummer. 30240580
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240580 250 mg
30240775 100 mg
30240744 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240580 Leverandør Toronto Research Chemicals Leverandørnr. TRCA618635250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 192139-92-7
Molekylær formel C31H35ClN2O2RuS
Molekylvægt (g/mol) 636.21
Synonym [R-(R*,R*)]-[N-[2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium, [N-[(1R,2R)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium
SMIL CC(C)c1ccc(C)cc1.Cc2ccc(cc2)S(=O)(=O)N([Ru]Cl)[C@@H]([C@H](N)c3ccccc3)c4ccccc4
Tekniske data
Kemisk navn eller materiale [((R,R)-2-Amino-1,2-diphenylethyl)[(4-tolyl)sulfonyl]amido](chloro)(eta6-p-cymene)ruthenium
Analyt- eller komponentnavne [((R,R)-2-Amino-1,2-diphenylethyl)[(4-tolyl)sulfonyl]amido](chloro)(eta6-p-cymene)ruthenium
Forsendelsestilstand Room Temperature
Løsningstype Neat
Industry Type Pharmaceutical