Få mere at vide
(R)-Quizalofop-tefuryl, TRC
Artikelnummer. 30138218
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138218 100 mg
30134997 25 mg
30004510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138218 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ826105100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 200509-41-7
Molekylær formel C22 H21 Cl N2 O5
Molekylvægt (g/mol) 428.87
Synonym C 4874, Pantera, Pantera 45EC, Quizalofop P tefuryl, Quizalofop-P-tefuryl, UBI-C 4874, Propanoic acid, 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-, (tetrahydro-2-furanyl)methyl ester, (2R)- (9CI, ACI)
IUPAC navn oxolan-2-ylmethyl (2R)-2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
SMIL C[C@@H](Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC4CCCO4
Tekniske data
Kemisk navn eller materiale Pantera
Mængde 100 mg
InChI formel InChI=1S/C22H21ClN2O5/c1-14(22(26)28-13-18-3-2-10-27-18)29-16-5-7-17(8-6-16)30-21-12-24-20-11-15(23)4-9-19(20)25-21/h4-9,11-12,14,18H,2-3,10,13H2,1H3/t14-,18?/m1/s1
Formel vægt 428.1139