Få mere at vide
(R)-Prunasin Tetraacetate, TRC
Artikelnummer. 30411380
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411380 10 mg
30411215 25 mg
30418277 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30411380 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83902510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60981-47-7
Molekylær formel C22H25NO10
Molekylvægt (g/mol) 463.43
Synonym (R)-alpha-[(2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl)oxy]benzeneacetonitrile
IUPAC navn [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[cyano(phenyl)methoxy]oxan-2-yl]methyl acetate
SMIL CC(=O)OC[C@H]1O[C@@H](OC(C#N)c2ccccc2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale (R)-Prunasin Tetraacetate
Mængde 10 mg
InChI formel InChI=1S/C22H25NO10/c1-12(24)28-11-18-19(29-13(2)25)20(30-14(3)26)21(31-15(4)27)22(33-18)32-17(10-23)16-8-6-5-7-9-16/h5-9,17-22H,11H2,1-4H3/t17?,18-,19-,20+,21-,22-/m1/s1
Formel vægt 463.1478