Få mere at vide
(R)-Propionyl Carnitine-d3 Chloride, TRC
Artikelnummer. 30413891
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30413891 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413891 Leverandør Toronto Research Chemicals Leverandørnr. TRCP78640225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Propionyl Carnitine-d3 Chloride
CAS 1334532-19-2
Molekylær formel C10 D3 H17 N O4 . Cl
Mængde 25 mg
Synonym 1-Propanaminium','3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxopropoxy)-','chloride (1:1)','(2R)- (ACI), [(2R)-4-Hydroxy-4-oxo-2-propanoyloxybutyl]-dimethyl-(trideuteriomethyl)azanium chloride, Levocarnitine-d3 propionate hydrochloride, L-Propionylcarnitine-d3, Propionyl-L-carnitine-d3 hydrochloride, Propionyl-L-carnitine-D3 (N-methyl-D3) hydrochloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxopropoxy)-1-propanaminium chloride, (2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium-d3 Chloride, (-)-Propionylcarnitine-d3, L-Carnitine-d3 Propionyl Ester, Dromos-d3, ST 261
InChI formel InChI=1S/C10H19NO4.ClH/c1-5-10(14)15-8(6-9(12)13)7-11(2,3)4;/h8H,5-7H2,1-4H3;1H/t8-;/m1./s1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CC
IUPAC navn [(2R)-3-carboxy-2-propanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 256.74
Formel vægt 256.1269