Få mere at vide
(R)-Pregabalin, TRC
Artikelnummer. 30138383
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138383 10 mg
17952575 50 mg
17964885 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138383 Leverandør Toronto Research Chemicals Leverandørnr. TRCP70478510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148553-51-9
Molekylær formel C8 H17 N O2
Molekylvægt (g/mol) 159.23
Synonym Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3R)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (R)-, (R)-3-(Aminomethyl)-5-methylhexanoic acid, (R)-Pregabalin, PD 144550, ent-Pregabalin
IUPAC navn (3R)-3-(aminomethyl)-5-methylhexanoic acid
SMIL CC(C)C[C@@H](CN)CC(=O)O
Tekniske data
Kemisk navn eller materiale (R)-Pregabalin
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/t7-/m1/s1
Mængde 10 mg
Formel vægt 159.1259
Procent renhed >95%