Få mere at vide
(R)-Pregabalin, Mikromol™
Artikelnummer. 17960766
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17960766 25 mg
17942766 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17960766 Leverandør Mikromol Leverandørnr. MM1376.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(R)-Pregabalin
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Pregabalin
CAS 148553-51-9
Analyt- eller komponentnavne (R)-Pregabalin
Forsendelsestilstand Room Temperature
Molekylær formel C8 H17 N O2
Certificeringer/compliance ISO 17025
Synonym Pregabalin Imp. B (EP), Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3R)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (R)-, (R)-3-(Aminomethyl)-5-methylhexanoic acid, (R)-Pregabalin, PD 144550, ent-Pregabalin, Pregabalin Related Compound A, Pregabalin Impurity B
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/t7-/m1/s1
SMIL CC(C)C[C@@H](CN)CC(=O)O
IUPAC navn (3R)-3-(aminomethyl)-5-methylhexanoic acid
Molekylvægt (g/mol) 159.23
Mængde 25 mg
Formel vægt 159.1259 g/mol