Få mere at vide
(R)-Pramipexole-d3 Dihydrochloride, TRC
Artikelnummer. 30416356
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416356 1 mg
30418165 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416356 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7007471MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1432230-09-5
Molekylær formel C10H16D3Cl2N3S
Molekylvægt (g/mol) 287.27
Synonym (R)-4,5,6,7-Tetrahydro-N6-(propyl-d3)-2,6-benzothiazolediamine Dihydrochloride, R-(+)-Pramipexole-d3 Dihydrochloride, Dexpramipexole-d3 Dihydrochloride
SMIL NC(S1)=NC2=C1C[C@H](NCCC([2H])([2H])[2H])CC2
Tekniske data
Kemisk navn eller materiale (R)-Pramipexole-d3 Dihydrochloride
Mængde 1 mg
InChI formel InChI=1S/C10H17N3S/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8/h7,12H,2-6H2,1H3,(H2,11,13)/t7-/m1/s1/i1D3
Formel vægt 286.0865