Få mere at vide
(R)-Ofloxacin-d3, TRC
Artikelnummer. 30410482
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30410482 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410482 Leverandør Toronto Research Chemicals Leverandørnr. TRCO24574225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Ofloxacin-d3
CAS 1346617-10-4
Molekylær formel C18 2H3 H17 F N3 O4
Mængde 25 mg
Synonym (3R)-9-Fluoro-2,3-dihydro-3-methyl-10-[4-(methyl-d3)-1-piperazinyl]-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, (R)-Ofloxacin-D3 (N-methyl-D3), (R)-Ofloxacin-d3
InChI formel InChI=1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/t10-/m1/s1/i2D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)c2c(F)cc3C(=O)C(=CN4[C@H](C)COc2c34)C(=O)O
Molekylvægt (g/mol) 364.39
Formel vægt 364.1626