Få mere at vide
R-(-)-O-Desmethyl Venlafaxine, TRC
Artikelnummer. 30354645
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354645 50 mg
30349797 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354645 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29650550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142761-11-3
Molekylær formel C16 H25 N O2
Molekylvægt (g/mol) 263.38
Synonym (R)-O-Desmethylvenlafaxine, Phenol, 4-[(1R)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, (R)-, 4-[(1R)-2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, (-)-O-Desmethylvenlafaxine, (R)-(-)-O-Desmethylvenlafaxine, R-(-)-O-Desmethyl venlafaxine
IUPAC navn 4-[(1R)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
SMIL CN(C)C[C@@H](c1ccc(O)cc1)C2(O)CCCCC2
Tekniske data
Kemisk navn eller materiale R-(-)-O-Desmethyl Venlafaxine
Mængde 50 mg
InChI formel InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/t15-/m0/s1
Formel vægt 263.1885