Få mere at vide
(R)-(-)-Norlaudanosine Hydrochloride, TRC
Artikelnummer. 30410188
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30410188 500 mg
1 options
Artikelnummer. 30410188 Leverandør Toronto Research Chemicals Leverandørnr. TRCN661415500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-(-)-Norlaudanosine Hydrochloride
CAS 54417-53-7
Molekylær formel C20H26ClNO4
Mængde 500 mg
Synonym (R)-1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, D)-(-)-Norlaudanosine Hydrochloride, (R)-Tetrahydropapaverine Hydrochloride, D-(-)-Tetrahydropapaverine Hydrochloride
InChI formel InChI=1S/C20H25NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-6,10-12,16,21H,7-9H2,1-4H3;1H/t16-;/m1./s1
SMIL Cl.COc1ccc(C[C@H]2NCCc3cc(OC)c(OC)cc23)cc1OC
IUPAC navn (1R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
Molekylvægt (g/mol) 379.88
Formel vægt 379.155