Få mere at vide
(R)-Norfluoxetine-d5 Phthalimide(Phenyl-d5), TRC
Artikelnummer. 30426639
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30426639 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426639 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68114225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Norfluoxetine-d5 Phthalimide(Phenyl-d5)
CAS 1346617-46-6
Molekylær formel C24H13D5F3NO3
Mængde 25 mg
Synonym 2-[(3R)-3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-1H-isoindole-1,3(2H)-dione-d5, (R)-1-(Phthalimido)-3-[(4-trifluoromethylphenyl)oxy]-3-phenylpropane-d5
InChI formel InChI=1S/C24H18F3NO3/c25-24(26,27)17-10-12-18(13-11-17)31-21(16-6-2-1-3-7-16)14-15-28-22(29)19-8-4-5-9-20(19)23(28)30/h1-13,21H,14-15H2/t21-/m1/s1/i1D,2D,3D,6D,7D
IUPAC navn (R)-2-(3-(phenyl-d5)-3-(4-(trifluoromethyl)phenoxy)propyl)isoindoline-1,3-dione
Molekylvægt (g/mol) 430.43
Formel vægt 430.16