Få mere at vide
(R)-N'-Nitrosonornicotine, TRC
Artikelnummer. 30466753
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466753 50 mg
30474659 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466753 Leverandør Toronto Research Chemicals Leverandørnr. TRCN53503050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61601-78-3
Molekylær formel C9H11N3O
Molekylvægt (g/mol) 177.2
Synonym 3-[(2R)-1-Nitroso-2-pyrrolidinyl]pyridine, (R)-3-(1-Nitroso-2-pyrrolidinyl)pyridine
SMIL [H][C@]1(C2=CC=CN=C2)CCCN1N=O
Tekniske data
Kemisk navn eller materiale (R)-N'-Nitrosonornicotine
Mængde 50 mg
InChI formel InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2/t9-/m1/s1
Formel vægt 177.0902