Få mere at vide
(R)-N-Methyl-Laudanosine Iodide, TRC
Artikelnummer. 30479666
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479666 500 mg
30474165 250 mg
30476185 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479666 Leverandør Toronto Research Chemicals Leverandørnr. TRCM315665500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 41431-32-7
Molekylær formel C22H30INO4
Molekylvægt (g/mol) 499.38
Synonym (R)-6,7-dimethoxy-2,2-dimethyliodide, (-)-Laudanosine Methiodide, (R)-N-Methyllaudanosinium Iodide, D-(-)-Laudanosine Methiodide
IUPAC navn (1R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2,2-dimethyl-3,4-dihydro-1H-isoquinolin-2-ium;iodide
SMIL [I-].COc1ccc(C[C@@H]2c3cc(OC)c(OC)cc3CC[N+]2(C)C)cc1OC
Tekniske data
Kemisk navn eller materiale (R)-N-Methyl-Laudanosine Iodide
Mængde 500 mg
InChI formel InChI=1S/C22H30NO4.HI/c1-23(2)10-9-16-13-21(26-5)22(27-6)14-17(16)18(23)11-15-7-8-19(24-3)20(12-15)25-4;/h7-8,12-14,18H,9-11H2,1-6H3;1H/q+1;/p-1/t18-;/m1./s1
Formel vægt 499.122