missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-4-Chlorophenylalanine, 95%, 98% ee, Thermo Scientific™

Catalog Number p-69778
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57292-44-1
C14H18ClNO4
299.75
MFCD00076978
BETBOAZCLSJOBQ-LLVKDONJSA-N
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
7018791
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O
This item is not returnable. View return policy
57292-44-1
C14H18ClNO4
299.75
MFCD00076978
BETBOAZCLSJOBQ-LLVKDONJSA-N
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
7018791
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 110°C
Color White
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 299.75
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-BOC-4-Chlorophenylalanine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'