missing translation for 'onlineSavingsMsg'
Learn More

(R)-Methotrimeprazine, TRC

Product Code. 30470253
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30470253 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470253 Supplier Toronto Research Chemicals Supplier No. TRCM2607701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Levomepromazine
CAS 60-99-1
Molekylær formel C19 H24 N2 O S
Mængde 1 mg
Synonym 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N,β-trimethyl-, (βR)-, 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N,β-trimethyl-, (R)-, Phenothiazine, 10-[3-(dimethylamino)-2-methylpropyl]-2-methoxy-, (-)- (8CI), (βR)-2-Methoxy-N,N,β-trimethyl-10H-phenothiazine-10-propanamine, 2-Methoxytrimeprazine, 7044 R.P., CL 36467, CL 39743, Hirnamin, Levomeprazine, Levomepromazin, Levomepromazine, Levopromazine, Levoprome, Levopromethazine, Levotomin, Mepromazine, Methotrimeprazine, NSC 226516, Neozine, Nirvan, Nozinan, Nozinane, RP 7044, SKF 5116, Sinogan, Sinogan-Debil, Tisercin, XP 03
InChI formel InChI=1S/C19H24N2OS/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21/h5-11,14H,12-13H2,1-4H3/t14-/m1/s1
SMIL COc1ccc2Sc3ccccc3N(C[C@H](C)CN(C)C)c2c1
IUPAC navn (2R)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
Molekylvægt (g/mol) 328.47
Formel vægt 328.1609
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.