Få mere at vide
R-Mannostatin A, Hydrochloride (~90%), TRC
Artikelnummer. 30465374
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30465374 0.5 mg 0.5mg
30470207 5 mg -
30475527 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30465374 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1665550.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 134677-22-8
Molekylær formel C6H13NO3S . HCl
Molekylvægt (g/mol) 215.7
Synonym (1S,2S,3S,4R,5S)-4-Amino-5-(methylthio)cyclopentane-1,2,3-triol Hydrochloride
IUPAC navn (1S,2S,3S,4R,5S)-4-amino-5-methylsulfanylcyclopentane-1,2,3-triol;hydrochloride
SMIL Cl.CS[C@H]1[C@H](N)[C@H](O)[C@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale R-Mannostatin A, Hydrochloride
Mængde 0.5 mg
InChI formel InChI=1S/C6H13NO3S.ClH/c1-11-6-2(7)3(8)4(9)5(6)10;/h2-6,8-10H,7H2,1H3;1H/t2-,3+,4+,5+,6+;/m1./s1
Formel vægt 215.0383