Få mere at vide
(R)-Lercanidipine-d3 Hydrochloride, TRC
Artikelnummer. 30470613
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30470613 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470613 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1790121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Lercanidipine-d3 Hydrochloride
CAS 1217724-52-1
Molekylær formel C36H39D3ClN3O6
Mængde 1 mg
Synonym (4R)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)(methyl-d3)amino]-1,1-dimethylethyl Methyl Ester Hydrochloride
InChI formel InChI=1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H/t33-;/m1./s1/i5D3;
SMIL O=C(OC(C)(C)CN(C([2H])([2H])[2H])CCC(C1=CC=CC=C1)C2=CC=CC=C2)C3=C(C)NC(C)=C(C(OC)=O)[C@H]3C4=CC([N+]([O-])=O)=CC=C4.Cl
Molekylvægt (g/mol) 651.21