Få mere at vide
R&D Systems™ Human SLC6A4/5-HTTLPR (NP_001036) VersaClone cDNA
Artikelnummer. 18758438
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18758438 10 μg
1 options
Artikelnummer. 18758438 Leverandør R&D Systems™ Leverandørnr. RDC1060
Solute Carrier Family 6 Member 4
The Human SLC6A4/5-HTTLPR (NP_001036) VersaClone cDNA clone contains the complete ORF for SLC6A4/5-HTTLPR/Serotonin transporter, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC6A4/5-HTTLPR/Serotonin transporter insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Mængde 10 μg
For Reasearch Use Only; Not for Use in Humans