Få mere at vide
R&D Systems™ Human Connexin 30/GJB6 (NP_006774) VersaClone cDNA
Artikelnummer. 18726479
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18726479 10 μg
1 options
Artikelnummer. 18726479 Leverandør R&D Systems™ Leverandørnr. RDC1101
CX30; DFNA3B; DFNB1B; ED2; EDH; GJB6
The Human Connexin 30/GJB6 (NP_006774) VersaClone cDNA clone contains the complete ORF for Connexin 30/GJB6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 30/GJB6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Mængde 10 μg
For Reasearch Use Only; Not for Use in Humans