Få mere at vide
R&D Systems™ Human CEACAM-20 (NP_001096067) VersaClone cDNA
Artikelnummer. 18739008
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18739008 10 μg
1 options
Artikelnummer. 18739008 Leverandør R&D Systems™ Leverandørnr. RDC1043
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 14-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Carcinoembryonic Antigen-related Cell Adhesion Molecule 20
The Human CEACAM-20 (NP_001096067) VersaClone cDNA clone contains the complete ORF for CEACAM20, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM20 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Tekniske data
Gene Alias CEACAM20, GPAD9366, UNQ9366
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Mængde 10 μg
For Reasearch Use Only; Not for Use in Humans