Få mere at vide
(R)-Haloxyfop Methyl Ester, TRC
Artikelnummer. 30137892
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137892 1 g
30133049 250 mg
17961235 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137892 Leverandør Toronto Research Chemicals Leverandørnr. TRCH1040201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72619-32-0
Molekylær formel C16 H13 Cl F3 N O4
Molekylvægt (g/mol) 375.73
Synonym Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-, methyl ester, (R)-, (R)-Haloxyfop methyl ester, (R)-Haloxyfop-methyl, DE 535, Edge, Eloge, Gallant super, Haloxyfop-P-methyl, Haloxyfop-R-methyl, Zellek Super
IUPAC navn methyl (2R)-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
SMIL COC(=O)[C@@H](C)Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1
Tekniske data
Kemisk navn eller materiale Haloxyfop-R-methyl
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/t9-/m1/s1
Formel vægt 375.0485
Procent renhed >95%