Få mere at vide
(R)-Duloxetine Hydrochloride, TRC
Artikelnummer. 30379138
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30379138 100 mg
1 options
Artikelnummer. 30379138 Leverandør Toronto Research Chemicals Leverandørnr. TRCD720990100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Duloxetine Hydrochloride
CAS 910138-96-4
Molekylær formel C18 H19 N O S . Cl H
Mængde 100 mg
Synonym 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride (1:1), (γR)-, 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride, (γR)- (9CI), (R)-Duloxetine hydrochloride, (R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine hydrochloride, Duloxetine Hydrochloride Imp. A (EP) as Hydrochloride
InChI formel InChI=1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m1./s1
SMIL Cl.CNCC[C@@H](Oc1cccc2ccccc12)c3cccs3
IUPAC navn (3R)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-2-ylpropan-1-amine;hydrochloride
Molekylvægt (g/mol) 333.88
Formel vægt 333.0954