Få mere at vide
Tocris Bioscience™ XL 413 Hydrochloride
Potent and selective Cdc7 inhibitor
Brand: Tocris Bioscience™ 5493/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
XL 413 hydrochloride is a potent and selective Cdc7 inhibitor (IC50 = 3.4 nM). Exhibits >12-fold selectivity for Cdc7 over PIM-1 kinase and >30-fold selectivity over pMCM and CK2. Inhibits proliferation of Colo 205 cells in vitro and attenuates tumor growth of Colo 205 xenografts in mice.Tekniske data
| 8-Chloro-2-(2S)-2-pyrrolidinylbenzofuro[3,2-d]pyrimidin-4(3H)-one hydrochloride | |
| 1169562-71-3 | |
| 50 mg | |
| Soluble to 20mM in water |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only