Få mere at vide
Tocris Bioscience™ Vancomycin HCl
Antibiotic
Brand:  Tocris Bioscience™ 5506/1G
Encompass
Produktkode 15716725
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Vancomycin HCl is an antibiotic with activity against Gram-positive bacteria.
Tekniske data
Tekniske data
Vancomycin HCl
1 g
Solid
1404-93-9
Soluble to 25mM in water
Bioactive Small Molecule
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use