Få mere at vide
Tocris Bioscience™ TC-I 2014
Beskrivelse
TC-I 2014 is a potent TRPM8 antagonist (IC50 values are 0.8, 3.0 and 4.4 nM for canine, human and rat channels respectively). Exhibits antiallodynic properties in pain models in vivo.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | TC-I 2014 |
| CAS | 1221349-53-6 |
| Synonym | 3-[7-(Trifluoromethyl)-5-[2-(trifluoromethyl)phenyl]-1H-benzimidazol-2-yl]-1-oxa-2-azaspiro[4.5]dec-2-ene |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molekylær formel | C23H19F6N3O |
| Formel vægt | 467.41 |
| Fysisk form | Powder |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
Research Use Only. Not For Human or Veterinary Use