Få mere at vide
Tocris Bioscience™ TC-G 1005
Beskrivelse
TC-G 1005 is a potent and selective GPBA agonist (EC50 values are 0.72 nM and 6.2 nM for hTGR5 and mTGR5, respectively). TC-G 1005 is selective for TGR5 over FXR (farnesoid X receptor). TC-G 1005 increases plasma GLP-1 levels and reduces blood glucose in mice. Orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | TC-G 1005 |
| CAS | 1415407-60-1 |
| Synonym | (4-Cyclopropyl-3,4-dihydro-1(2H)-quinoxalinyl)[4-(2,5-dimethylphenoxy)-3-pyridinyl]methanone |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molekylær formel | C25H25N3O2 |
| Formel vægt | 399.48 |
| Fysisk form | Solid |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
Research Use Only. Not For Human or Veterinary Use