Få mere at vide
R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X)
Optimized for the extended culture of pluripotent stem cell-derived cardiomyocytes. R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X) is nutrient-rich, fully defined, and serum-free.
Brand:  R&D Systems™ AR011
Encompass
Produktkode 15720013
Advarsel:
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
  • Maintains healthy cardiomyocyte population for longer than 200 days
  • High lot-to-lot consistency decreases experimental variation
Repeated mechanical stress can negatively affect cell attachment and survival. For extended culture, daily media exchanges should be handled as gently as possible.
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.