Få mere at vide
R&D Systems™ Rat IL-2 ELISpot Kit
Artikelnummer. 18772174
Skift visning
Mængde:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18772174 1 x 96-well Plate
18732434 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18772174 Leverandør R&D Systems™ Leverandørnr. EL502
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
TRUSTED_SUSTAINABILITY
Tekniske data
Inkluderer one 96-well PVDF-backed plate coated w/pAb for rat IL-2, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Mængde 1 x 96-well Plate
Prøvetype Whole Cells
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Til brug med (applikation) Diagnostics
Produkttype Rat IL-2 ELISpot Kit