Få mere at vide
R&D Systems™ Rat IFN-gamma ELISpot Kit
Artikelnummer. 18782254
Skift visning
Mængde:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18782254 1 x 96-well Plate
18712304 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18782254 Leverandør R&D Systems™ Leverandørnr. EL585
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
TRUSTED_SUSTAINABILITY
Tekniske data
Inkluderer One 96-well PVDF-backed plate coated w/pAb for rat IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT; Positive control
Mængde 1 x 96-well Plate
Prøvetype Whole Cells
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Til brug med (applikation) Diagnostics
Produkttype Rat IFN-gamma ELISpot Kit