Learn More
R&D Systems™ Mouse Relaxin R1 (NP_997617) VersaClone cDNA
Relaxin Receptor 1
Brand: R&D Systems™ RDC0790
This item is not returnable.
View return policy
Description
The Mouse Relaxin R1 (NP_997617) VersaClone cDNA clone contains the complete ORF for Relaxin R1/LGR7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Relaxin R1/LGR7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mRXFP1 | |
10 ÎĽg at 0.2 ÎĽg/ÎĽL | |
2289bp | |
Mouse |
NP_997617 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 ÎĽg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse Relaxin R1 (NP_997617) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction