Få mere at vide
R&D Systems™ Mouse Mesenchymal Stem Cell Functional Identification Kit
Artikelnummer. 18761814
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
18761814 1 sæt
1 options
Artikelnummer. 18761814 Leverandør R&D Systems™ Leverandørnr. SC010
 Begrænset vare
Forventet forsendelse: 20-11-2026
Indeholder
Adipogenic Supplement, Mouse/Rat Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP4, Sheep Anti-mouse Collagen II, Goat Anti-mouse Osteopontin
To verify multipotency of mouse mesenchymal stem cells by in vitro functional differentiation
Reduces experimental variation
Reliably induces MSC trilineage differentiation using defined supplements
A panel of antibodies, consisting of Goat Anti-mouse FABP4, Sheep Anti-mouse Collagen II, and Goat Anti-mouse Osteopontin, are included to define th
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Produktlinje R&D Systems
Mængde 1 sæt