missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Mouse GPR171 (NP_775574) VersaClone cDNA
Beskrivelse
The Mouse GPR171 (NP_775574) VersaClone cDNA clone contains the complete ORF for GPR171, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR171 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature.
Tekniske data
Tekniske data
| Gene | mGPR171 |
| Adgangsnummer | NP_775574 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Indsæt type | 972bp |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Arter | Mouse |
| Mængde | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?