missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Mouse CRTH-2 (NP_034092) VersaClone cDNA

Artikelnummer. 18471570
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18471570 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18471570 Leverandør R&D Systems™ Leverandørnr. RDC0116
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 13-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Vea la política de devoluciones

Chemoattractant Receptor-homologous Molecule Expressed on Th2 Cells

The Mouse CRTH-2 (NP_034092) VersaClone cDNA clone contains the complete ORF for CRTH-2/GPR44, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CRTH-2/GPR44 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Información de órdenes

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Especificaciones

Gene mCRTH2
Adgangsnummer NP_034092
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.5
Indsæt type 1161bp
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Mouse
Mængde 10 μg

For Reasearch Use Only; Not for Use in Humans

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.