Få mere at vide
Tocris Bioscience™ (-)-JQ1
Inactive control for (+)-JQ1
Brand: Tocris Bioscience™ 5603/10
Beskrivelse
(-)-JQ1 is an inactive control for (+)-JQ1 . Exhibits no significant interaction with BRD1-4 or a panel of other bromodomains.Active Enantiomer also available.
Tekniske data
| (-)-JQ1 | |
| (6R)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetic acid 1,1-dimethylethyl ester | |
| Soluble to 100mM in DMSO and to 100mM in ethanol | |
| Bioactive Small Molecule |
| 1268524-71-5 | |
| 10 mg | |
| Solid |
Research Use Only. Not For Human or Veterinary Use