missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TCIRG1 (NP_006010) VersaClone cDNA

Product Code. 18452320
missing translation for 'orderingAttributeHoverText'
Mængde:
10 μg
This item is not returnable. View return policy

Product Code. 18452320

missing translation for 'mfr': R&D Systems™ RDC0186

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

T-cell Immune Regulator 1

The Human TCIRG1 (NP_006010) VersaClone cDNA clone contains the complete ORF for TCIRG1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TCIRG1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Gene hTCIRG1
Adgangsnummer NP_006010
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.5
Indsæt type 2506bp
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Human
Mængde 10 μg

For Reasearch Use Only; Not for Use in Humans

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.