missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-23R (AAM44229) VersaClone cDNA
IL-23R; interleukin 23 receptor; interleukin-23 receptor
Brand: R&D Systems™ RDC1255
This item is not returnable.
View return policy
Description
The Human IL-23 R (AAM44229) VersaClone cDNA clone contains the complete ORF for IL-23R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-23R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature

Specifications
cDNA | |
AAM44229 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
149233, 209590 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
hIL23R | |
10 μg at 0.2 μg/μL | |
1930bp | |
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
Human, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human IL-23R (AAM44229) VersaClone cDNA > 10μg
For Reasearch Use Only; Not for Use in Humans.
Spot an opportunity for improvement?Share a Content Correction