Få mere at vide
R&D Systems™ Human IL-13 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 213-ILB-01MG/CF
Encompass
Produktkode 15719883
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
The Recombinant Human IL-13 Protein is derived from E. coli. The Recombinant Human IL-13 Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
Lyophilized from a 0.2μm filtered solution in PBS.
12.3 kDa
1mg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
3596
IL-13
E. coli-derived human IL-13 protein Gly21-Asn132
Unconjugated
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
R&D Systems™ Human IL-13 Recombinant Protein