Få mere at vide
R&D Systems™ Human Coagulation Factor II/Thrombin Protein
Mængde:
200 μg
Beskrivelse
The Human Coagulation Factor II/Thrombin Protein is derived from Human Plasma. The Human Coagulation Factor II/Thrombin Protein has been validated for the following applications: Enzyme Activity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in MES, NaCl and Brij-35. |
| Gen-id (Entrez) | 2147 |
| Navn | Coagulation Factor II/Thrombin |
| Mængde | 200 μg |
| Kilde | Human plasma-derived Coagulation Factor II/Thrombin protein The human plasma used for the isolation of this product was certified by the supplier to be non-reactive for HIV-1/2 and HBsAg negative at the donor level. Human blood products should always be treated in accordance with universal handling precautions. |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturel form | pro form |
| Konjugeret | Unconjugated |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |