missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Human BTNL2/Butyrophilin-like 2 (NP_062548) VersaClone cDNA

Artikelnummer. 18461410
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18461410 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18461410 Leverandør R&D Systems™ Leverandørnr. RDC0823
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 13-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Butyrophilin-like 2

The Human BTNL2/Butyrophilin-like 2 (NP_062548) VersaClone cDNA clone contains the complete ORF for BTNL2/Butyrophilin-like 2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The BTNL2/Butyrophilin-like 2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Ordreinfo

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Tekniske data

Gene BTNL2/Butyrophilin-like 2
Adgangsnummer NP_062548
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.5
Indsæt type 1381bp
Gen-id (Entrez) 56244
Gene Alias BTL-II, Butyrophilin like 2, Butyrophilin-like 2, butyrophilin-like 2 (MHC class II associated)
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Human
Mængde 10 μg

For Reasearch Use Only; Not for Use in Humans

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.