Få mere at vide
Tocris Bioscience™ EPZ 004777
Beskrivelse
EPZ 004777 is a highly potent DOT1L inhibitor (IC50 = 0.4 nM). Exhibits >1000-fold selectivity for DOT1L over a panel of other methyltransferases. Selectively inhibits proliferation and induces apoptosis of MLL-rearranged cells in vitro. Prolongs survival in a MLL xenograft mouse model. Enhances efficiency of Yamanaka-factor factor reprogramming of fibroblasts by 3-4-fold.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | EPZ 004777 |
| CAS | 1338466-77-5 |
| Synonym | 7-[5-Deoxy-5-[[3-[[[[4-(1,1-dimethylethyl)phenyl]amino]carbonyl]amino]propyl](1-methylethyl)amino]-β-D-ribofuranosyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molekylær formel | C28H41N7O4 |
| Formel vægt | 539.67 |
| Fysisk form | Solid |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
Research Use Only. Not For Human or Veterinary Use