Få mere at vide
Tocris Bioscience™ Edaglitazone
Beskrivelse
Edaglitazone is a potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Edaglitazone |
| CAS | 213411-83-7 |
| Synonym | 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione |
| Mængde | 50 mg |
| Opløselighed | Soluble to 100mM in DMSO |
| Molekylær formel | C24H20N2O4S2 |
| Formel vægt | 464.56 |
| Fysisk form | Solid |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
Research Use Only. Not For Human or Veterinary Use