Få mere at vide
R&D Systems™ Caspase-4 Inhibitor Z-YVAD-FMK
Mængde:
1 mg
Beskrivelse
- Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
- Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
- Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
Tekniske data
Tekniske data
| Indhold og opbevaring | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Mængde | 1 mg |
| Produkttype | Caspase-4 |
| Molekylvægt (g/mol) | 630Da |
| Inhibitorer | Z-YVAD-FMK |
| Form | Lyophilized |
For research use only. Not for use in diagnostic procedures