Få mere at vide
R&D Systems™ Canine IL-2 ELISpot Kit
Tekniske data
Tekniske data
| Inkluderer | One 96-well plate coated w/polyclonal Ab for canine IL-2, Biotinylated detection Ab, Streptavidin conjugated to AP, Buffers, BCIP/NBT chromogen, Positive control |
| Mængde | 1 x 96-well Plate |
| Prøvetype | Whole Cells |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Til brug med (applikation) | Diagnostics |
| Produkttype | Canine IL-2 ELISpot Kit |