Få mere at vide
R&D Systems™ Boc-VPR-AMC Fluorogenic Peptide Substrate
Mængde:
20 mg
Beskrivelse
- Substrate for Coagulation Factor II, Coagulation Factor VII, Kallikrein 4, Kallikrein 5, Kallikrein 8, Kallikrein 13, Kallikrein 14, and MASP3
- Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
- A fluorescence plate reader with excitation at 380nmand emission at 460nmis recommended for the measurement of the enzymatic activity
- Purity >95% based on TLC
Tekniske data
Tekniske data
| Mængde | 20 mg |
| Produkttype | Boc-Val-Pro-Arg-AMC |
| Opbevaringskrav | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |