Få mere at vide
R&D Systems™ Boc-QAR-AMC Fluorogenic Peptide Substrate
Mængde:
10 mg
Beskrivelse
- Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
- A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
- Purity >98%
- Excellent substrate for several serine proeases
Tekniske data
Tekniske data
| Mængde | 10 mg |
| Produkttype | Boc-Gln-Ala-Arg-AMC HCl |
| Opbevaringskrav | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for three months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |