Få mere at vide
(R)-Crizotinib-d5 (piperidine-3,3,4,5,5-d5), TRC
Artikelnummer. 30350456
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30350456 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350456 Leverandør Toronto Research Chemicals Leverandørnr. TRCC7850015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Crizotinib-d5 (piperidine-3,3,4,5,5-d5)
CAS 1395950-84-1
Molekylær formel C21 D5 H17 Cl2 F N5 O
Mængde 5 mg
Synonym 2-Pyridinamine, 3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl-3,3,4,5,5-d5)-1H-pyrazol-4-yl]- (ACI), 3-[(1R)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl-3,3,4,5,5-d5)-1H-pyrazol-4-yl]-2-pyridinamine (ACI)
InChI formel InChI=1S/C21H22Cl2FN5O/c1-12(19-16(22)2-3-17(24)20(19)23)30-18-8-13(9-27-21(18)25)14-10-28-29(11-14)15-4-6-26-7-5-15/h2-3,8-12,15,26H,4-7H2,1H3,(H2,25,27)/t12-/m1/s1/i4D2,5D2,15D
SMIL [2H]C1([2H])CNCC([2H])([2H])C1([2H])n2cc(cn2)c3cnc(N)c(O[C@H](C)c4c(Cl)ccc(F)c4Cl)c3
IUPAC navn 3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(3,3,4,5,5-pentadeuteriopiperidin-4-yl)pyrazol-4-yl]pyridin-2-amine
Molekylvægt (g/mol) 455.3675
Formel vægt 454.1499