Få mere at vide
(R)-Citalopram Oxalate, Mikromol™
Artikelnummer. 17972226
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17972226 25 mg
1 options
Artikelnummer. 17972226 Leverandør Mikromol Leverandørnr. MM1162.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(R)-Citalopram Oxalate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-Citalopram Oxalate
CAS 219861-53-7
Analyt- eller komponentnavne (R)-Citalopram Oxalate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H21 F N2 O . C2 H2 O4
Certificeringer/compliance ISO 17025
Synonym (R)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile ethanedioate, R-(-)-Citalopram oxalate, Orlistat Related Compound A (USP), 1-(3-Dimethylaminopropyl)-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile Oxalate, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, (1R)-, ethanedioate (1:1), (R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile ethanedioate, Escitalopram Oxalate Impurity K as Oxalate, Escitalopram Impurity K as Oxalate
InChI formel InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m1./s1
SMIL CN(C)CCC[C@@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O
IUPAC navn (1R)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molekylvægt (g/mol) 414.43
Mængde 25 mg
Formel vægt 414.1591 g/mol