Få mere at vide
Tocris Bioscience™ (R)-Citalopram oxalate
GREENER_CHOICE
Enantiomer of escitalopram oxalate (Cat. No. 1427)
Brand:  Tocris Bioscience™ 5763/50
Encompass
Produktkode 15757096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
(R)-Citalopram oxalate is an enantiomer of escitalopram oxalate . Postulated to allosterically modulate the affinity of the S-enantiomer.
Racemate and S-enantiomer also available.
Tekniske data
Tekniske data
(-)-(R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate
219861-53-7
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner