Få mere at vide
(R)-(+)-Celiprolol-d9 Hydrochloride, TRC
Artikelnummer. 30363688
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363688 5 mg
30358999 10 mg
30358214 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363688 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2544925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1330045-90-3
Molekylær formel C20H24D9N3O4 • HCl
Molekylvægt (g/mol) 425.02
Synonym N'-[3-Acetyl-4-[(2R)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethylurea-d9 Hydrochloride, (+)-(R)-Celiprolol-d9 Hydrochloride, (+)-Celiprolol-d9 Hydrochloride, (R)-Celiprolol-d9 Hydrochloride, d-Celiprolol-d9 Hydrochloride
IUPAC navn (R)-3-(3-acetyl-4-(2-hydroxy-3-((2-(methyl-d3)propan-2-yl-1,1,1,3,3,3-d6)amino)propoxy)phenyl)-1,1-diethylurea hydrogen chloride
SMIL CCN(CC)C(NC1=CC(C(C)=O)=C(OC[C@H2]([OH])CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1)=O.Cl
Tekniske data
Kemisk navn eller materiale (R)-(+)-Celiprolol-d9 Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H/t16-;/m1./s1/i4D3,5D3,6D3;
Formel vægt 424.02