Få mere at vide
(R)-(+)-Camphor-d2, TRC
Artikelnummer. 30339077
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30339077 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339077 Leverandør Toronto Research Chemicals Leverandørnr. TRCC16580210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-(+)-Camphor-d2
CAS 34733-73-8
Molekylær formel C10H14D2O
Mængde 10 mg
Synonym (1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one-3,3-d2
InChI formel InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10-/m1/s1/i6D2
SMIL [2H]C1([2H])[C@H]2CC[C@](C)(C1=O)C2(C)C
IUPAC navn (1R,4S)-2,2-dideuterio-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
Molekylvægt (g/mol) 154.25
Formel vægt 154.1327